Anti-GRINA Rabbit Polyclonal Antibody

Lieferant: US Biological
USBI036306EA 587 EUR
USBI036306
Anti-GRINA Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: GRINA (glutamate receptor, ionotropic, N-methyl D-aspartate-associated protein 1 (glutamate binding))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR