Anti-FABP7 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: US Biological
USBI035334-APCEA 850 EUR
USBI035334-APC
Anti-FABP7 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Do not freeze!

Type: Primary
Antigen: FABP7 (fatty acid binding protein 7, brain)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR