Anti-DTL Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI034834EA 627 EUR
USBI034834
Anti-DTL Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50

Type: Primary
Antigen: DTL (denticleless E3 ubiquitin protein ligase homolog (Drosophila))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR