Anti-DDIT3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Lieferant: US Biological
USBI034535-APCEA 852 EUR
USBI034535-APC
Anti-DDIT3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: DDIT3 (DNA-damage-inducible transcript 3)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human