Anti-VSTM4 Rabbit Polyclonal Antibody

Lieferant: US Biological
USBI032647EA 587 EUR
USBI032647
Anti-VSTM4 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000; Flow Cytometry: 1:10-50

Type: Primary
Antigen: VSTM4 (V-set and transmembrane domain containing 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR