Anti-APOA2 Rabbit Polyclonal Antibody

Lieferant: US Biological
USBI031976EA 587 EUR
USBI031976
Anti-APOA2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:10-50

Type: Primary
Antigen: APOA2 (apolipoprotein A-II)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR