Anti-ANKRD39 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
USBI031921-BIOTINEA 791 EUR
USBI031921-BIOTIN
Anti-ANKRD39 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100

Type: Primary
Antigen: ANKRD39 (ankyrin repeat domain 39)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR