Anti-ATG4D Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 222CT15.4.1]

Lieferant: United States Biological
USBI030795-FITCEA 852 EUR
USBI030795-FITC
Anti-ATG4D Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 222CT15.4.1]
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1000; Immunofluorescence: 1:10-50

Type: Primary
Antigen: ATG4D (autophagy related 4D, cysteine peptidase)
Clonality: Monoclonal
Clone: 222CT15.4.1
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR