Anti-CRTC3 Mouse Monoclonal Antibody [clone: 13B196]
Lieferant: United States Biological
USBI030118EA 590 EUR
USBI030118
Anti-CRTC3 Mouse Monoclonal Antibody [clone: 13B196]
Antikörper
Type: Primary
Antigen: CRTC3 (CREB regulated transcription coactivator 3)
Clonality: Monoclonal
Clone: 13B196
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey