Anti-IgG Goat Polyclonal Antibody (CF™ 640R)

Lieferant: Biotium

20081-1EA 144 EUR
20081-1 20081.
Anti-IgG Goat Polyclonal Antibody (CF™ 640R)
Antikörper
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF® 640R Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR