Anti-TRMT1L Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB121883-100EA 816 EUR
ABCAAB121883-100
Anti-TRMT1L Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TRMT1L (tRNA methyltransferase 1 homolog (S. cerevisiae)-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR