Anti-KIAA1045 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB121643-100EA 816 EUR
ABCAAB121643-100
Anti-KIAA1045 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KIAA1045 (KIAA1045)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human