Anti-RAPH1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB121619-100EA 816 EUR
ABCAAB121619-100
Anti-RAPH1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RAPH1 (Ras association (RalGDS/AF-6) and pleckstrin homology domains 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR