Anti-EQTN Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB121470-100EA 816 EUR
ABCAAB121470-100
Anti-EQTN Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EQTN (equatorin, sperm acrosome associated)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR