Anti-KIAA0513 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB121430-100EA 816 EUR
ABCAAB121430-100
Anti-KIAA0513 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KIAA0513 (KIAA0513)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human