Anti-IGSF9B Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB121324-100EA 816 EUR
ABCAAB121324-100
Anti-IGSF9B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IGSF9B (immunoglobulin superfamily, member 9B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR