Anti-GAB3 Rabbit polyclonal antibody unconjugated
Lieferant: Abcam
ABCAAB121311-100EA 816 EUR
ABCAAB121311-100
Anti-GAB3 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: GAB3 (GRB2-associated binding protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human