Anti-OST-beta Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB121285-100EA 839 EUR
ABCAAB121285-100
Anti-OST-beta Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: OST-beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR