Anti-IFT43 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB121195-100EA 816 EUR
ABCAAB121195-100
Anti-IFT43 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IFT43 (intraflagellar transport 43 homolog (Chlamydomonas))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR