Anti-Factor D Mouse Monoclonal Antibody [clone: D10/4]

Lieferant: Abcam
ABCAAB121054-100EA 842 EUR
ABCAAB121054-100
Anti-Factor D Mouse Monoclonal Antibody [clone: D10/4]
Antikörper
Type: Primary
Antigen: Factor D
Clonality: monoclonal
Clone: D10/4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR