Anti-Influenza A Virus Hemagglutinin Mouse Monoclonal Antibody [clone: 1.B.408]

Lieferant: Abcam
ABCAAB119966-100EA 840 EUR
ABCAAB119966-100
Anti-Influenza A Virus Hemagglutinin Mouse Monoclonal Antibody [clone: 1.B.408]
Antikörper
Type: Primary
Antigen: Influenza A Virus Hemagglutinin
Clonality: monoclonal
Clone: 1.B.408
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR