Anti-FCGR1A Mouse monoclonal antibody unconjugated [clone: 10.1]
Lieferant: Abcam
ABCAAB119843-100EA 862 EUR
ABCAAB119843-100
Anti-FCGR1A Mouse monoclonal antibody unconjugated [clone: 10.1]
Antikörper
Type: Primary
Antigen: FCGR1A (Fc fragment of IgG, high affinity Ia, receptor (CD64))
Clonality: monoclonal
Clone: 10.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen