Anti-Glutamate Receptor 1 (AMPA subtype) Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB119397-100EA 840 EUR
ABCAAB119397-100
Anti-Glutamate Receptor 1 (AMPA subtype) Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: Glutamate Receptor 1 (AMPA subtype)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR