Anti-EFHD2 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB119119-50EA 835 EUR
ABCAAB119119-50
Anti-EFHD2 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: EFHD2 (EF-hand domain family, member D2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR