Anti-PANK2 Mouse monoclonal antibody [clone: 3H9]
Lieferant: Abcam
ABCAAB119070-100EA 755 EUR
ABCAAB119070-100
Anti-PANK2 Mouse monoclonal antibody [clone: 3H9]
Type: Primary
Antigen: PANK2 (pantothenate kinase 2)
Clonality: monoclonal
Clone: 3H9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen