Anti-ITM2B Mouse monoclonal antibody [clone: 1C11]
Lieferant: Abcam
ABCAAB119044-100EA 708 EUR
ABCAAB119044-100
Anti-ITM2B Mouse monoclonal antibody [clone: 1C11]
Type: Primary
Antigen: ITM2B (integral membrane protein 2B)
Clonality: monoclonal
Clone: 1C11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human