Anti-SLC39A2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB118619-100EA 859 EUR
ABCAAB118619-100
Anti-SLC39A2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SLC39A2 (solute carrier family 39 (zinc transporter), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR