Anti-Josephin-1 Rabbit polyclonal antibody

Lieferant: Abcam
ABCAAB118221-100EA 724 EUR
ABCAAB118221-100
Anti-Josephin-1 Rabbit polyclonal antibody
Type: Primary
Antigen: Josephin-1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR