Anti-SSH2 Rabbit polyclonal antibody

Lieferant: Abcam
ABCAAB118079-100EA 744 EUR
ABCAAB118079-100
Anti-SSH2 Rabbit polyclonal antibody
Type: Primary
Antigen: SSH2 (slingshot protein phosphatase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR