Anti-GALE Mouse monoclonal antibody [clone: 1C4]

Lieferant: Abcam
ABCAAB118033-100EA 755 EUR
ABCAAB118033-100
Anti-GALE Mouse monoclonal antibody [clone: 1C4]
Type: Primary
Antigen: GALE (UDP-galactose-4-epimerase)
Clonality: monoclonal
Clone: 1C4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR