Anti-AK5 Mouse monoclonal antibody [clone: 1D6]

Lieferant: Abcam
ABCAAB117927-100EA 772 EUR
ABCAAB117927-100
Anti-AK5 Mouse monoclonal antibody [clone: 1D6]
Type: Primary
Antigen: AK5 (adenylate kinase 5)
Clonality: monoclonal
Clone: 1D6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR