Anti-SH3BP2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB117833-100EA 837 EUR
ABCAAB117833-100
Anti-SH3BP2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SH3BP2 (SH3-domain binding protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR