Anti-ANKHD1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB117788-100EA 837 EUR
ABCAAB117788-100
Anti-ANKHD1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ANKHD1 (ankyrin repeat and KH domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR