Anti-MYC Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 9E10]

Lieferant: Abcam
ABCAAB117599-100EA 834 EUR
ABCAAB117599-100
Anti-MYC Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 9E10]
Antikörper
Type: Primary
Antigen: MYC (v-myc avian myelocytomatosis viral oncogene homolog)
Clonality: monoclonal
Clone: 90000000000
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR