Anti-Estradiol Mouse Monoclonal Antibody [clone: AB4.2]

Lieferant: Abcam
ABCAAB116648-100EA 949 EUR
ABCAAB116648-100
Anti-Estradiol Mouse Monoclonal Antibody [clone: AB4.2]
Antikörper
Type: Primary
Antigen: Estradiol
Clonality: monoclonal
Clone: AB4.2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR