Anti-Cortisol Mouse Monoclonal Antibody [clone: F4P1A3]

Lieferant: Abcam
ABCAAB116600-100EA 835 EUR
ABCAAB116600-100
Anti-Cortisol Mouse Monoclonal Antibody [clone: F4P1A3]
Antikörper
Type: Primary
Antigen: Cortisol
Clonality: monoclonal
Clone: F4P1A3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR