Anti-AVPR1B Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB116246-50EA 797 EUR
ABCAAB116246-50
Anti-AVPR1B Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AVPR1B (arginine vasopressin receptor 1B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR