Anti-TSLP Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB115700-50EA 862 EUR
ABCAAB115700-50
Anti-TSLP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TSLP (thymic stromal lymphopoietin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR