Anti-PARP2 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB115620-50EA 826 EUR
ABCAAB115620-50
Anti-PARP2 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PARP2 (poly (ADP-ribose) polymerase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR