Anti-CysLT2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB115453-50EA 800 EUR
ABCAAB115453-50
Anti-CysLT2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CysLT2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR