Anti-LGI4 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB115305-50EA 797 EUR
ABCAAB115305-50
Anti-LGI4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LGI4 (leucine-rich repeat LGI family, member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR