Anti-RLF Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB115154-100EA 797 EUR
ABCAAB115154-100
Anti-RLF Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RLF (rearranged L-myc fusion)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR