Anti-delta Sarcoglycan Goat polyclonal antibody unconjugated
Lieferant: Abcam
ABCAAB115019-200EA 840 EUR
ABCAAB115019-200
Anti-delta Sarcoglycan Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: delta Sarcoglycan
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Rat