Anti-IRF2BP2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB114997-100EA 840 EUR
ABCAAB114997-100
Anti-IRF2BP2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IRF2BP2 (interferon regulatory factor 2 binding protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR