Anti-SEPT3 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB113983-50EA 797 EUR
ABCAAB113983-50
Anti-SEPT3 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SEPT3 (septin 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR