Anti-CD40 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB113701-100EA 823 EUR
ABCAAB113701-100
Anti-CD40 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CD40 (CD40 molecule, TNF receptor superfamily member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR