Anti-NAMPT Rabbit Polyclonal Antibody (Biotin)

Lieferant: Abcam
ABCAAB113615-50EA 835 EUR
ABCAAB113615-50
Anti-NAMPT Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: NAMPT (nicotinamide phosphoribosyltransferase)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR