Anti-LY9 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB113285-100EA 859 EUR
ABCAAB113285-100
Anti-LY9 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LY9 (lymphocyte antigen 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR