Anti-ACTB Rabbit Polyclonal Antibody (Biotin)

Lieferant: Abcam
ABCAAB113279-100EA 840 EUR
ABCAAB113279-100
Anti-ACTB Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: ACTB (actin, beta)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR