Anti-TRMT12 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB113103-100EA 797 EUR
ABCAAB113103-100
Anti-TRMT12 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TRMT12 (tRNA methyltransferase 12 homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR